Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc18a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153054.2 | NP_694694.1 | 12 | 1522 | ||
GenBank mRNA | AK076433.1 | - | 11 | 1247 | ||
AY779336.1 | - | 10 | 1180 | |||
BC027192.1 | - | 12 | 1460 | 4239930 | ||
BC031146.1 | - | 12 | 1490 | 4974776 |