Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pygb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153781.1 | NP_722476.1 | 20 | 3260 | ||
GenBank mRNA | AK032769.1 | - | 20 | 3273 | ||
AK035673.1 | - | 20 | 2990 | |||
AK133794.1 | - | 11 | 1990 | |||
AK134667.1 | - | 20 | 3270 | |||
AK142716.1 | - | 20 | 3270 | |||
AK147763.1 | - | 20 | 3292 | |||
AK168381.1 | - | 20 | 3273 | |||
AK169052.1 | - | 20 | 3295 | |||
AK169631.1 | - | 20 | 3272 | |||
BC031457.1 | - | 20 | 3267 | |||
BC032209.1 | - | 14 | 2415 | |||
BC035283.1 | - | 20 | 3260 | 4947455 |