Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AKR1A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001202413.1 | NP_001189342.1 | 8, 9 | 1123 | ||
NM_001202414.1 | NP_001189343.1 | 9, 10 | 1251 | |||
NM_006066.3 | NP_006057.1 | 8, 9 | 1340 | |||
NM_153326.2 | NP_697021.1 | 7, 8 | 1212 | |||
XM_011540491.2 | XP_011538793.1 | 6, 7 | 1071 | |||
XM_011540492.2 | XP_011538794.1 | 7, 8 | 1037 | |||
GenBank mRNA | AK293083.1 | - | 7, 8 | 948 | ||
BC000670.2 | - | 8, 9 | 1269 | 3349300 | ||
BC005394.1 | - | 7, 8 | 928 | 4051944 | ||
BT007003.1 | - | 6, 7 | 833 | |||
CR457010.1 | - | 6, 7 | 833 | |||
EU668319.1 | - | 7, 8 | 1169 | |||
EU794588.1 | - | 7, 8 | 1169 | |||
J04794.1 | - | 7, 8 | 893 |