Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Thsd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040426.3 | NP_001035516.2 | 9 | 1730 | ||
NM_172444.4 | NP_766032.1 | 3 | 538 | |||
GenBank mRNA | AK045414.1 | - | 3 | 454 | ||
BC139329.1 | - | 3 | 431 | 8862351 | ||
CV559893.1 | - | 3 | 538 |