Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Galnt6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161767.1 | NP_001155239.1 | 12 | 3113 | ||
NM_001161768.1 | NP_001155240.1 | 12 | 3110 | |||
NM_172451.3 | NP_766039.2 | 12 | 3109 | |||
GenBank mRNA | AK028506.1 | - | 12 | 3109 | ||
AK155008.1 | - | 12 | 3116 | |||
AK159721.1 | - | 12 | 3091 |