Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Srsf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001078167.2 | NP_001071635.1 | 1 | 540 | ||
NM_173374.4 | NP_775550.2 | 1 | 540 | |||
GenBank mRNA | AK018176.1 | - | 1 | 209 | ||
AK053674.1 | - | 1 | 230 | |||
AK078715.1 | - | 1 | 197 | |||
AK150535.1 | - | 1 | 243 | |||
AK169203.1 | - | 1 | 196 | |||
AK169623.1 | - | 1 | 198 | |||
AK181394.1 | - | 1 | 46 | |||
BC046773.1 | - | 1 | 172 | 5708288 | ||
BC058627.1 | - | 1 | 206 | 6401674 | ||
BY743476.1 | - | 1 | 243 | |||
CF739274.1 | - | 1 | 540 | |||
X66091.1 | - | 1 | 153 |