Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbxw17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175401.3 | NP_780610.2 | 10 | 1415 | ||
GenBank mRNA | AK075676.1 | - | 2 | 1540 | ||
AK135553.1 | - | 8 | 2267 | |||
AK155157.1 | - | 11 | 1929 | |||
BC040428.1 | - | 10 | 1415 | 3327345 |