Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HAX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006118.3 | NP_006109.2 | 2 | 245 | ||
GenBank mRNA | AK290626.1 | - | 2 | 167 | ||
AK294298.1 | - | 2 | 171 | |||
AK310000.1 | - | 2 | 165 | |||
AK313291.1 | - | 2 | 179 | |||
BC005240.1 | - | 2 | 297 | 3931805 | ||
BC014314.1 | - | 2 | 166 | 3931852 | ||
BC015209.1 | - | 2 | 138 | 3883802 | ||
BC016730.1 | - | 2 | 183 | 4082810 | ||
EU190982.1 | - | 2 | 179 | |||
EU190983.1 | - | 2 | 162 | |||
KU178523.1 | - | 2 | 84 | |||
U68566.1 | - | 2 | 245 |