Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CAPZA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006135.2 | NP_006126.1 | 4, 5 | 605 | ||
XM_011542225.2 | XP_011540527.1 | 4, 5 | 349 | |||
XM_017002424.1 | XP_016857913.1 | 4, 5 | 349 | |||
GenBank mRNA | AK223226.1 | - | 4, 5 | 226 | ||
AK289654.1 | - | 4, 5 | 248 | |||
BC000144.1 | - | 4, 5 | 230 | 3047937 | ||
BT019364.1 | - | 4, 5 | 213 | |||
BX648738.1 | - | 4, 5 | 896 | |||
CR407657.1 | - | 4, 5 | 213 | |||
CR541819.1 | - | 4, 5 | 213 | |||
U56637.1 | - | 4, 5 | 213 |