Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dcdc2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195617.1 | NP_001182546.1 | 4, 5 | 832 | ||
NM_177577.3 | NP_808245.2 | 4, 5 | 832 | |||
GenBank mRNA | AK220370.1 | - | 4, 5 | 605 | ||
BC045136.1 | - | 4, 5 | 832 | 6306251 |