Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Setd1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040398.2 | NP_001035488.2 | 10 | 3665 | ||
GenBank mRNA | AK122435.1 | - | 2 | 123 | ||
BC035291.1 | - | 2 | 167 | |||
BC038367.1 | - | 4 | 743 | |||
BC040775.1 | - | 2 | 167 | |||
BC041681.1 | - | 2 | 186 |