Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abhd16a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178592.3 | NP_848707.1 | 5 | 439 | ||
GenBank mRNA | BC029114.1 | - | 5 | 439 | 2648618 |