Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tbc1d10c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178650.3 | NP_848765.2 | 10 | 1222 | ||
GenBank mRNA | AK038366.1 | - | 2 | 362 | ||
AK040487.1 | - | 10 | 1223 | |||
AK075803.1 | - | 10 | 1209 | |||
AK088119.1 | - | 1217 | ||||
AK131177.1 | - | 9 | 1152 | |||
AK146426.1 | - | 10 | 1216 | |||
AK153836.1 | - | 10 | 1221 | |||
AK154444.1 | - | 9 | 1613 | |||
BC094901.1 | - | 9 | 1197 |