Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bend7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190400.1 | NP_001177329.1 | 8, 9 | 1649 | ||
NM_178663.4 | NP_848778.1 | 8, 9 | 1652 | |||
GenBank mRNA | AK030428.1 | - | 8, 9 | 1339 | ||
AK053896.1 | - | 8, 9 | 1651 | |||
BC056476.1 | - | 5, 6 | 696 | |||
BC096430.1 | - | 8, 9 | 1321 | 5041388 | ||
BC119370.1 | - | 8, 9 | 1291 | 8734119 | ||
BC144864.1 | - | 8, 9 | 1288 | 9053412 |