Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Me3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181407.4 | NP_852072.2 | 9, 10 | 1217 | ||
GenBank mRNA | AK006146.1 | - | 4, 5 | 373 | ||
AK032196.1 | - | 9, 10 | 1218 | |||
AK080798.1 | - | 9, 10 | 1150 | |||
BC099935.1 | - | 9, 10 | 1313 | 30097608 |