Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Esrp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290383.1 | NP_001277312.1 | 16 | 3457 | ||
NM_194055.3 | NP_918944.2 | 16 | 3460 | |||
GenBank mRNA | BC031468.1 | - | 12 | 2490 | 4037840 | |
BC059280.1 | - | 12 | 2500 | |||
DV648780.1 | - | 1 | 347 |