Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc9c1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198106.4 | NP_932774.3 | 4 | 538 | ||
GenBank mRNA | AY368685.1 | - | 4 | 498 | ||
BC145031.1 | - | 243 | ||||
BC145032.1 | - | 243 |