Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wwox | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019573.3 | NP_062519.2 | 3 | 338 | ||
GenBank mRNA | AF187014.1 | - | 3 | 289 | ||
AK018507.1 | - | 3 | 268 | |||
AK019911.1 | - | 3 | 296 | |||
AK046903.1 | - | 3 | 288 | |||
AK078528.1 | - | 3 | 291 | |||
AK083096.1 | - | 3 | 270 | |||
BC014716.1 | - | 3 | 294 | 4241066 | ||
CJ058964.1 | - | 3 | 339 |