Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Paqr4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023824.3 | NP_076313.1 | 3 | 2408 | ||
GenBank mRNA | AK005142.1 | - | 3 | 2431 | ||
AY424293.1 | - | 3 | 2359 | |||
BC011185.1 | - | 3 | 2414 | 4196033 | ||
BC026921.1 | - | 3 | 2359 | 4500871 |