Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arfgap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166024.1 | NP_001159496.1 | 17 | 2496 | ||
NM_023854.2 | NP_076343.2 | 16 | 2454 | |||
GenBank mRNA | AF229439.1 | - | 16 | 2263 | ||
AK009565.1 | - | 17 | 2320 | |||
AK141749.1 | - | 4 | 930 | |||
AK149837.1 | - | 16 | 2289 | |||
AK168489.1 | - | 16 | 2264 | |||
AK169144.1 | - | 16 | 2277 | |||
AK209318.1 | - | 1 | 135 | |||
BC005495.1 | - | 16 | 2302 | 3487811 |