Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Elp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023876.3 | NP_076365.2 | 5 | 593 | ||
GenBank mRNA | AJ276004.1 | - | 5 | 592 | ||
AK020742.1 | - | 5 | 601 | |||
AK034040.1 | - | 5 | 598 | |||
AK034529.1 | - | 5 | 599 | |||
AK037104.1 | - | 5 | 617 | |||
AK039447.1 | - | 5 | 605 | |||
AK039758.1 | - | 5 | 594 | |||
AK079069.1 | - | 5 | 608 | |||
AK168964.1 | - | 5 | 657 |