Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tsc22d4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023910.6 | NP_076399.4 | 5 | 2100 | ||
GenBank mRNA | AF201286.1 | - | 5 | 1813 | ||
AK018735.2 | - | 5 | 2073 | |||
AK040534.1 | - | 5 | 2037 | |||
AK143608.1 | - | 5 | 1877 | |||
AK150043.1 | - | 5 | 2069 | |||
AK154153.1 | - | 5 | 2050 | |||
AK154490.1 | - | 5 | 1700 | |||
BC018544.1 | - | 4 | 1572 | 3666221 | ||
BC023761.1 | - | 5 | 2004 | 5343927 |