Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sun1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256115.1 | NP_001243044.1 | 3 | 221 | ||
NM_001256116.1 | NP_001243045.1 | 2 | 158 | |||
NM_001256117.1 | NP_001243046.1 | 3 | 221 | |||
NM_001256118.1 | NP_001243047.1 | 2 | 158 | |||
NM_024451.2 | NP_077771.1 | 3 | 221 | |||
GenBank mRNA | AK014585.1 | - | 3 | 219 | ||
AK036187.1 | - | 3 | 198 | |||
AK160281.1 | - | 3 | 179 | |||
AK161959.1 | - | 3 | 193 | |||
AK162533.1 | - | 3 | 192 | |||
AK167686.1 | - | 3 | 193 | |||
AK196775.1 | - | 2 | 92 | |||
AK198340.1 | - | 2 | 92 | |||
AK200268.1 | - | 2 | 92 | |||
AY682989.1 | - | 2 | 138 | |||
BC047928.1 | - | 3 | 188 | 5321879 | ||
BC048156.1 | - | 3 | 190 | 5698402 | ||
BQ551727.1 | - | 2 | 162 | |||
BY036585.1 | - | 3 | 221 | |||
CK389844.1 | - | 3 | 221 | |||
HQ402597.1 | - | 2 | 138 |