Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sun1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256115.1 | NP_001243044.1 | 7 | 884 | ||
NM_024451.2 | NP_077771.1 | 7 | 884 | |||
GenBank mRNA | AK036187.1 | - | 7 | 861 | ||
AK160281.1 | - | 7 | 842 | |||
AK167686.1 | - | 7 | 856 | |||
BC047928.1 | - | 7 | 851 | 5321879 | ||
BC048156.1 | - | 7 | 853 | 5698402 | ||
HQ141858.1 | - | 2 | 265 | |||
HQ141859.1 | - | 2 | 265 | |||
HQ141860.1 | - | 2 | 265 | |||
HQ141861.1 | - | 2 | 265 | |||
HQ141862.1 | - | 2 | 265 | |||
HQ141863.1 | - | 2 | 265 | |||
HQ141864.1 | - | 2 | 265 | |||
HQ141865.1 | - | 2 | 265 | |||
HQ141866.1 | - | 2 | 265 |