Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lias | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024471.4 | NP_077791.1 | 2 | 208 | ||
GenBank mRNA | AB057731.1 | - | 2 | 107 | ||
AK013575.1 | - | 1 | 707 | |||
AK031851.1 | - | 2 | 180 | |||
AK048744.1 | - | 152 | ||||
AK049548.1 | - | 2 | 123 | |||
AK077192.1 | - | 2 | 209 | |||
BC002141.1 | - | 2 | 140 | 3484559 |