Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sh3bp5l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK086088.1 | - | 7 | 2076 | ||
AK122543.1 | - | 5 | 5280 | |||
AK144928.1 | - | 7 | 2075 | |||
AK150129.1 | - | 7 | 2055 | |||
AK156171.1 | - | 7 | 2116 | |||
AK159202.1 | - | 7 | 2107 | |||
AK166491.1 | - | 7 | 2076 | |||
AK167203.1 | - | 7 | 2193 | |||
AK172333.1 | - | 7 | 2116 | |||
BC003251.1 | - | 6 | 1737 | 3500047 | ||
BC013661.1 | - | 3 | 1208 | |||
BC094418.1 | - | 7 | 2098 | 30702568 |