Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spsb3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163750.1 | NP_001157222.1 | 7 | 1533 | ||
NM_001163751.1 | NP_001157223.1 | 7 | 1188 | |||
NM_027141.2 | NP_081417.2 | 7 | 1209 | |||
GenBank mRNA | AF403038.1 | - | 7 | 1209 | ||
AK009325.1 | - | 7 | 1209 | |||
AK076170.1 | - | 7 | 1541 | |||
AK082768.1 | - | 7 | 1132 | |||
AK156579.1 | - | 6 | 1546 | |||
AK167474.1 | - | 6 | 1526 | |||
AK220234.1 | - | 7 | 1471 | |||
BC026523.1 | - | 7 | 1874 | |||
BC072556.1 | - | 7 | 1494 | 5691114 |