Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atg2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029654.4 | NP_083930.5 | 36 | 5693 | ||
GenBank mRNA | AK220468.1 | - | 1 | 3317 | ||
BC024533.1 | - | 4 | 315 | |||
BC046427.1 | - | 15 | 1804 | |||
BC062182.1 | - | 36 | 5663 | 6505402 |