Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAD21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006265.2 | NP_006256.1 | 2, 3 | 440 | ||
GenBank mRNA | AK097915.1 | - | 2, 3 | 248 | ||
AK289505.1 | - | 2, 3 | 217 | |||
BC050381.1 | - | 2, 3 | 280 | 6044010 | ||
D38551.1 | - | 2, 3 | 336 | |||
DA328186.1 | - | 2, 3 | 440 | |||
X98294.1 | - | 1, 2 | 152 |