Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnft1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029788.5 | NP_084064.1 | 9 | 2557 | ||
GenBank mRNA | AK036401.1 | - | 9 | 2557 |