Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arsk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029847.4 | NP_084123.2 | 4 | 678 | ||
GenBank mRNA | AK013194.2 | - | 4 | 630 | ||
AK019515.1 | - | 4 | 644 | |||
AK031147.1 | - | 4 | 666 | |||
AK032812.1 | - | 4 | 644 | |||
AK033968.1 | - | 4 | 679 | |||
AK035464.1 | - | 4 | 599 | |||
AK039765.1 | - | 4 | 674 | |||
AK046378.1 | - | 4 | 679 | |||
AK083188.1 | - | 4 | 632 | |||
AK084090.1 | - | 4 | 664 | |||
AK086667.1 | - | 4 | 637 | |||
AK139856.1 | - | 2 | 218 | |||
AK144817.1 | - | 4 | 646 | |||
AK165970.1 | - | 4 | 646 | |||
BC046790.1 | - | 4 | 652 | 5717223 | ||
BC058351.1 | - | 4 | 652 | 5717223 |