Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NDC80 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006101.2 | NP_006092.1 | 4 | 433 | ||
GenBank mRNA | AF017790.1 | - | 4 | 355 | ||
AK289396.1 | - | 4 | 387 | |||
AK313184.1 | - | 4 | 385 | |||
BC005239.1 | - | 4 | 403 | |||
BC010171.2 | - | 4 | 361 | |||
BC035617.1 | - | 4 | 364 | 5164424 |