Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem50b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030018.3 | NP_084294.1 | 7 | 980 | ||
GenBank mRNA | AK020977.1 | - | 7 | 985 | ||
AK098154.1 | - | 7 | 944 | |||
AK133966.1 | - | 7 | 1008 | |||
AK150773.1 | - | 968 | ||||
AK151796.1 | - | 7 | 1010 | |||
AK152190.1 | - | 7 | 996 | |||
AK152767.1 | - | 7 | 996 | |||
AK155082.1 | - | 7 | 1033 | |||
AK170684.1 | - | 7 | 975 | |||
AK172335.1 | - | 7 | 968 | |||
BC004841.1 | - | 7 | 983 | 2649264 |