Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cstad | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030137.2 | NP_084413.1 | 2 | 104 | ||
GenBank mRNA | AK020589.1 | - | 2 | 106 | ||
AK047294.1 | - | 2 | 105 | |||
BC117017.1 | - | 2 | 100 | 40126336 | ||
BC117019.1 | - | 2 | 100 | 40126338 |