Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gins3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030198.3 | NP_084474.1 | 3 | 1898 | ||
GenBank mRNA | AK019225.1 | - | 3 | 1899 | ||
AK146574.1 | - | 3 | 1930 | |||
AK160606.1 | - | 3 | 1899 | |||
BC064746.1 | - | 3 | 1887 | 30095310 |