Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abtb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030251.3 | NP_084527.2 | 4 | 409 | ||
GenBank mRNA | AB053477.1 | - | 4 | 340 | ||
AK049100.1 | - | 4 | 325 | |||
AK088437.1 | - | 4 | 374 | |||
AK089474.1 | - | 2 | 773 | |||
AK155432.1 | - | 2 | 1372 | |||
AK155505.1 | - | 2 | 1372 | |||
AK155644.1 | - | 2 | 1372 | |||
AK220208.1 | - | 4 | 266 | |||
BC003234.1 | - | 4 | 353 | 3483893 | ||
BY201769.1 | - | 4 | 367 |