Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trim34b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243916.1 | NP_001230845.1 | 3, 4 | 832 | ||
GenBank mRNA | BC145515.1 | - | 3, 4 | 832 |
Gene Symbol: Trim34a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030684.3 | NP_109609.2 | 4, 5 | 1091 | ||
GenBank mRNA | AF220139.1 | - | 4, 5 | 988 | ||
AF220140.1 | - | 991 | ||||
AK089187.1 | - | 1106 | ||||
AK171840.1 | - | 4, 5 | 1051 | |||
AK172135.1 | - | 4, 5 | 1051 |