Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smarcd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031842.2 | NP_114030.2 | 4 | 531 | ||
GenBank mRNA | BC026783.1 | - | 4 | 443 | 4481097 | |
BC059921.1 | - | 4 | 501 | |||
BC079839.1 | - | 4 | 477 | 6810462 | ||
CV562378.1 | - | 4 | 531 | |||
M25773.1 | - | 4 | 487 | |||
U66620.1 | - | 4 | 487 |