Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dqx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033606.3 | NP_291084.1 | 6 | 1459 | ||
GenBank mRNA | AF318278.1 | - | 5 | 1103 | ||
BC048936.1 | - | 1 | 122 | |||
BC075680.1 | - | 6 | 1459 | 30612145 |