Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ophn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_052976.3 | NP_443208.1 | 7 | 1144 | ||
GenBank mRNA | AK028822.1 | - | 7 | 628 | ||
AK031419.1 | - | 7 | 1137 | |||
AK036038.1 | - | 7 | 1145 | |||
AK039304.1 | - | 6 | 1312 | |||
AK048153.1 | - | 7 | 1145 | |||
AK082551.1 | - | 7 | 784 | |||
AK087469.1 | - | 7 | 608 | |||
BC004845.1 | - | 8 | 742 | 2650009 |