Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pcdhb3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053128.2 | NP_444358.2 | 1 | 1098 | ||
GenBank mRNA | AF326296.1 | - | 1 | 914 | ||
AK030570.1 | - | 1 | 1098 | |||
AY013785.1 | - | 1 | 914 | |||
BC079557.1 | - | 1 | 1012 | 30360109 |