Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nans | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053179.3 | NP_444409.1 | 3 | 407 | ||
GenBank mRNA | AB041263.1 | - | 3 | 447 | ||
AK076290.1 | - | 3 | 408 | |||
AK088912.1 | - | 3 | 417 | |||
AK156254.1 | - | 3 | 452 | |||
AK169211.1 | - | 3 | 408 | |||
BC003307.1 | - | 3 | 500 | 3154822 | ||
BC057977.1 | - | 3 | 381 | 5052339 |