Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Fam131a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133778.2 | NP_598539.1 | 6 | 2172 | ||
GenBank mRNA | AK019338.1 | - | 1 | 1205 | ||
AK049983.1 | - | 6 | 2173 | |||
BC003957.1 | - | 1 | 1506 | |||
BC092532.1 | - | 6 | 2203 | 6594874 |