Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kmo | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133809.1 | NP_598570.1 | 2, 3 | 150 | ||
GenBank mRNA | AK129011.1 | - | 163 | |||
AK133074.1 | - | 258 | ||||
AK158251.1 | - | 2, 3 | 345 | |||
AK171812.1 | - | 2, 3 | 274 | |||
BC014683.1 | - | 2, 3 | 150 | 4210278 |