Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hnrnpf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166427.1 | NP_001159899.1 | 3 | 1325 | ||
NM_001166428.1 | NP_001159900.1 | 4 | 1264 | |||
NM_001166429.1 | NP_001159901.1 | 4 | 1225 | |||
NM_001166430.1 | NP_001159902.1 | 4 | 1576 | |||
NM_001166431.1 | NP_001159903.1 | 3 | 1515 | |||
NM_001166432.1 | NP_001159904.1 | 4 | 1316 | |||
NM_133834.2 | NP_598595.1 | 4 | 1207 | |||
GenBank mRNA | AK076478.1 | - | 3 | 1165 | ||
BC018185.1 | - | 3 | 1150 | 3995067 | ||
BC023162.1 | - | 1 | 231 | |||
BC025481.1 | - | 4 | 1184 | 5354000 | ||
BC027003.1 | - | 1 | 784 | 5365491 | ||
BC029163.1 | - | 1 | 968 | 4949890 | ||
BC029764.1 | - | 1 | 791 | 4947521 | ||
BC032197.1 | - | 4 | 1202 | |||
BC033483.1 | - | 1 | 966 | 4950131 | ||
BC042689.1 | - | 3 | 1448 | |||
BC047346.1 | - | 3 | 1448 | |||
BC089313.1 | - | 4 | 1049 | 6808998 |