Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HLA-DMA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006120.3 | NP_006111.2 | 3 | 649 | ||
GenBank mRNA | AK291858.1 | - | 3 | 585 | ||
AK298081.1 | - | 3 | 1227 | |||
BC011447.1 | - | 3 | 622 | 4245221 | ||
BC026279.1 | - | 3 | 578 | 4703588 | ||
CR450324.1 | - | 3 | 563 | |||
X62744.1 | - | 3 | 608 |