Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Usp53 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133857.3 | NP_598618.1 | 7 | 1287 | ||
GenBank mRNA | AK030317.1 | - | 8 | 1371 | ||
AK036876.1 | - | 7 | 1288 | |||
AK040332.1 | - | 8 | 1183 | |||
AK045953.1 | - | 7 | 1350 | |||
AK140453.1 | - | 7 | 1331 | |||
BC096383.1 | - | 7 | 1287 | |||
BC132339.1 | - | 6 | 1020 | 40130616 | ||
BC145707.1 | - | 6 | 1020 | 40131010 |