Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stx12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133887.4 | NP_598648.1 | 2 | 244 | ||
GenBank mRNA | AB019211.1 | - | 2 | 187 | ||
AK077678.1 | - | 2 | 245 | |||
AK078948.1 | - | 2 | 204 | |||
AK132618.1 | - | 2 | 208 | |||
AK146544.1 | - | 2 | 249 | |||
AK146735.1 | - | 2 | 236 | |||
AK167087.1 | - | 2 | 158 | |||
AK167804.1 | - | 2 | 208 | |||
AK168753.1 | - | 2 | 202 | |||
AK170203.1 | - | 2 | 281 | |||
AK187081.1 | - | 1 | 48 | |||
BC010669.1 | - | 2 | 231 | 2645010 |