Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Xpo1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001035226.1 | NP_001030303.1 | 3 | 286 | ||
NM_134014.3 | NP_598775.2 | 3 | 524 | |||
GenBank mRNA | BC029714.1 | - | 1 | 47 | ||
BC034710.1 | - | 2 | 280 | |||
BC062912.1 | - | 3 | 286 | 6821729 | ||
CA325110.1 | - | 3 | 286 | |||
CD347786.1 | - | 3 | 495 |