Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pi4k2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145501.2 | NP_663476.1 | 1 | 485 | ||
GenBank mRNA | AK030725.1 | - | 1 | 450 | ||
BC089309.1 | - | 1 | 426 | |||
BC110363.1 | - | 1 | 447 | 5346917 | ||
BY732177.1 | - | 1 | 486 |